siRNA Details
| Gene Symbol: Plce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019588.2 | NP_062534.2 | 1 | 283 | ||
| GenBank mRNA | AB076247.1 | - | 1 | 283 | ||
| BC025027.1 | - | 1 | 22 | |||
| BC138349.1 | - | 1 | 271 | 8861369 | ||
| BC138350.1 | - | 1 | 271 | 8861370 | ||
| BC145246.1 | - | 1 | 271 | 9053798 | ||

Plce1
Genomic Map