siRNA Details
| Gene Symbol: Smu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021535.4 | NP_067510.3 | 2 | 219 | ||
| GenBank mRNA | AB044414.1 | - | 2 | 148 | ||
| AK003493.1 | - | 2 | 206 | |||
| AK011140.1 | - | 2 | 218 | |||
| AK019174.1 | - | 2 | 178 | |||
| AK159635.1 | - | 2 | 206 | |||
| AK168813.1 | - | 2 | 213 | |||
| BC020034.1 | - | 2 | 161 | |||
| BC057446.1 | - | 2 | 180 | 3155760 | ||
| BY036103.1 | - | 2 | 221 | |||

Smu1
Genomic Map