siRNA Details
Gene Symbol: Slc29a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023596.3 | NP_076085.1 | 6 | 2438 | ||
GenBank mRNA | AK034133.1 | - | 6 | 2435 | ||
AK157868.1 | - | 5 | 2276 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc29a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023596.3 | NP_076085.1 | 6 | 2438 | ||
GenBank mRNA | AK034133.1 | - | 6 | 2435 | ||
AK157868.1 | - | 5 | 2276 |