siRNA Details
| Gene Symbol: Wdr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290521.1 | NP_001277450.1 | 9, 10 | 1091 | ||
| NM_001290522.1 | NP_001277451.1 | 9, 10 | 981 | |||
| NM_023597.2 | NP_076086.1 | 9, 10 | 984 | |||
| GenBank mRNA | AF274321.1 | - | 9, 10 | 961 | ||
| AK017259.1 | - | 9, 10 | 974 | |||
| AK141026.1 | - | 9, 10 | 1089 | |||
| BC026500.1 | - | 8, 9 | 870 | |||
| BC037102.1 | - | 9, 10 | 974 | 5364307 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001011976, XM_006238239, XM_006238237, BC079286.

Wdr31
Genomic Map