siRNA Details
Gene Symbol: Sars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023637.3 | NP_076126.2 | 1 | 182 | ||
GenBank mRNA | AB029949.1 | - | 1 | 154 | ||
AK010491.1 | - | 1 | 160 | |||
AK087606.1 | - | 1 | 149 | |||
AK135222.1 | - | 1 | 144 | |||
AK140760.1 | - | 1 | 149 | |||
BC079664.1 | - | 1 | 159 | 30357834 | ||
CX122368.1 | - | 1 | 182 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
