siRNA Details
| Gene Symbol: Trim29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023655.2 | NP_076144.2 | 2, 3 | 1012 | ||
| GenBank mRNA | AK013219.1 | - | 1, 2 | 63 | ||
| AK028448.1 | - | 2, 3 | 245 | |||
| BB612224.1 | - | 2, 3 | 242 | |||
| BC006699.1 | - | 2, 3 | 743 | |||
| BC027353.1 | - | 2, 3 | 927 | 3988926 | ||

Trim29
Genomic Map