siRNA Details
| Gene Symbol: Trim29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023655.2 | NP_076144.2 | 6, 7 | 1649 | ||
| GenBank mRNA | AF230390.1 | - | 4, 5 | 522 | ||
| AK013219.1 | - | 5, 6 | 700 | |||
| AK028448.1 | - | 6, 7 | 882 | |||
| BC006699.1 | - | 6, 7 | 1380 | |||
| BC027353.1 | - | 6, 7 | 1564 | 3988926 | ||

Trim29
Genomic Map