siRNA Details
| Gene Symbol: Spock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252620.1 | NP_001239549.1 | 5 | 365 | ||
| NM_001252621.1 | NP_001239550.1 | 4 | 356 | |||
| NM_023689.3 | NP_076178.1 | 5 | 479 | |||
| GenBank mRNA | AJ278998.1 | - | 5 | 373 | ||
| AK013644.1 | - | 5 | 408 | |||
| AK049843.1 | - | 4 | 357 | |||
| AK164338.1 | - | 5 | 353 | |||
| AK220280.1 | - | 5 | 463 | |||
| BC017601.1 | - | 5 | 378 | 4502169 | ||
| BC053334.1 | - | 5 | 350 | 6492113 | ||

Spock3
Genomic Map