siRNA Details
| Gene Symbol: Pi16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023734.3 | NP_076223.3 | 1 | 440 | ||
| GenBank mRNA | AB046537.1 | - | 89 | |||
| AK004672.1 | - | 1 | 134 | |||
| AK157854.1 | - | 1 | 446 | |||
| AK158339.1 | - | 1 | 442 | |||
| BC106088.1 | - | 1 | 125 | |||
| BC118026.1 | - | 1 | 88 | |||

Pi16
Genomic Map