siRNA Details
| Gene Symbol: 1700020D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023781.5 | NP_076270.3 | 1 | 650 | ||
| GenBank mRNA | AF319980.1 | - | 1 | 614 | ||
| AK006150.1 | - | 1 | 651 | |||
| BC049585.1 | - | 1 | 647 | 6772543 | ||
| BC061091.1 | - | 1 | 648 | |||

1700020D05Rik
Genomic Map