siRNA Details
| Gene Symbol: Fam13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143776.1 | NP_001137248.1 | 14 | 2004 | ||
| NM_001143777.1 | NP_001137249.1 | 15 | 2111 | |||
| NM_024244.4 | NP_077206.3 | 14 | 2007 | |||
| GenBank mRNA | AK004797.1 | - | 14 | 1991 | ||
| AK032053.1 | - | 16 | 1846 | |||
| AK041147.1 | - | 14 | 2005 | |||
| AK220421.1 | - | 14 | 3549 | |||
| BC003231.1 | - | 3 | 506 | |||
| BC066835.1 | - | 3 | 459 | |||
| BC129839.1 | - | 14 | 1960 | 40060810 | ||
| BC129840.1 | - | 14 | 1963 | 40060811 | ||

Fam13c
Genomic Map