siRNA Details
| Gene Symbol: Slc38a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164798.1 | NP_001158270.1 | 17 | 3027 | ||
| NM_001164799.1 | NP_001158271.1 | 16 | 3006 | |||
| NM_001164800.1 | NP_001158272.1 | 16 | 3003 | |||
| NM_024249.5 | NP_077211.4 | 17 | 3030 | |||
| GenBank mRNA | AK032245.1 | - | 17 | 2954 | ||
| AK159948.1 | - | 16 | 2968 | |||
| AK165672.1 | - | 16 | 2970 | |||
| AK188367.1 | - | 1 | 130 | |||
| BC005641.1 | - | 5 | 1379 | |||
| BC023375.1 | - | 1 | 159 | |||
| BC043299.1 | - | 7 | 1703 | |||
| BC055017.1 | - | 4 | 1051 | |||
| BC082300.1 | - | 16 | 2974 | 30362204 | ||
| BC088811.1 | - | 17 | 2963 | 6493546 | ||

Slc38a10
Genomic Map