siRNA Details
| Gene Symbol: Ccdc132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167750.1 | NP_001161222.1 | 7 | 594 | ||
| NM_001167751.1 | NP_001161223.1 | 7 | 594 | |||
| NM_024260.5 | NP_077222.4 | 7 | 594 | |||
| GenBank mRNA | AK042031.1 | - | 7 | 574 | ||
| AK075898.1 | - | 7 | 583 | |||
| AK129458.1 | - | 7 | 544 | |||
| AY254697.1 | - | 7 | 587 | |||
| BC036294.1 | - | 7 | 534 | 3500316 | ||
| BC043092.1 | - | 7 | 551 | 6405356 | ||

Ccdc132
Genomic Map