siRNA Details
| Gene Symbol: Ccdc132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167750.1 | NP_001161222.1 | 12 | 1056 | ||
| NM_001167751.1 | NP_001161223.1 | 12 | 1056 | |||
| NM_024260.5 | NP_077222.4 | 12 | 1056 | |||
| GenBank mRNA | AK042031.1 | - | 12 | 1036 | ||
| AK075898.1 | - | 12 | 1045 | |||
| AK129458.1 | - | 12 | 1006 | |||
| AY254697.1 | - | 12 | 1049 | |||
| BC036294.1 | - | 12 | 996 | 3500316 | ||
| BC043092.1 | - | 12 | 1013 | 6405356 | ||

Ccdc132
Genomic Map