siRNA Details
| Gene Symbol: Tsnaxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024445.4 | NP_077765.1 | 4, 5 | 414 | ||
| GenBank mRNA | AF343750.1 | - | 4, 5 | 386 | ||
| AK006041.1 | - | 4, 5 | 415 | |||
| BC100431.1 | - | 4, 5 | 427 | 30917958 | ||

Tsnaxip1
Genomic Map