siRNA Details
| Gene Symbol: Tsnaxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024445.4 | NP_077765.1 | 7 | 735 | ||
| GenBank mRNA | AF343750.1 | - | 7 | 707 | ||
| AK006041.1 | - | 7 | 736 | |||
| BC100431.1 | - | 7 | 748 | 30917958 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsnaxip1| Gene Symbol: Tsnaxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024445.4 | NP_077765.1 | 7 | 735 | ||
| GenBank mRNA | AF343750.1 | - | 7 | 707 | ||
| AK006041.1 | - | 7 | 736 | |||
| BC100431.1 | - | 7 | 748 | 30917958 | ||