siRNA Details
| Gene Symbol: Rnf219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026047.4 | NP_080323.3 | 6 | 806 | ||
| GenBank mRNA | BC029231.1 | - | 6 | 772 | 3156840 | |
| CN697474.1 | - | 3 | 331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf219| Gene Symbol: Rnf219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026047.4 | NP_080323.3 | 6 | 806 | ||
| GenBank mRNA | BC029231.1 | - | 6 | 772 | 3156840 | |
| CN697474.1 | - | 3 | 331 | |||