siRNA Details
| Gene Symbol: Wdr13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290783.1 | NP_001277712.1 | 9 | 3792 | ||
| NM_026137.5 | NP_080413.2 | 9 | 3826 | |||
| NM_028542.1 | NP_082818.1 | 9 | 4078 | |||
| GenBank mRNA | AK017549.1 | - | 9 | 3408 | ||
| AK075889.1 | - | 1 | 1721 | |||
| AK136979.1 | - | 1 | 1993 | |||
| BC047119.1 | - | 1 | 596 | |||
| BC057366.1 | - | 10 | 3576 | |||
| BC080853.1 | - | 9 | 3741 | |||
| BC108332.1 | - | 9 | 3582 | 6493226 | ||

Wdr13
Genomic Map