siRNA Details
| Gene Symbol: Snx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026998.3 | NP_081274.2 | 9 | 833 | ||
| GenBank mRNA | AK008135.1 | - | 4 | 298 | ||
| AK011640.1 | - | 10 | 835 | |||
| AK013158.1 | - | 9 | 834 | |||
| AK030764.1 | - | 3 | 163 | |||
| BC025911.1 | - | 9 | 820 | |||
| BC061028.1 | - | 10 | 874 | 6594585 | ||

Snx6
Genomic Map