siRNA Details
| Gene Symbol: 2310002L09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027104.3 | NP_081380.2 | 3 | 918 | ||
| GenBank mRNA | AK009097.1 | - | 3 | 919 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310002L09Rik| Gene Symbol: 2310002L09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027104.3 | NP_081380.2 | 3 | 918 | ||
| GenBank mRNA | AK009097.1 | - | 3 | 919 | ||