siRNA Details
| Gene Symbol: Atp6ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027439.4 | NP_081715.1 | 3 | 365 | ||
| GenBank mRNA | AB192471.2 | - | 3 | 241 | ||
| AK017482.1 | - | 3 | 297 | |||
| AK029405.1 | - | 3 | 268 | |||
| AK076495.1 | - | 3 | 310 | |||
| AK151735.1 | - | 3 | 296 | |||
| BC014706.1 | - | 3 | 236 | 4457079 | ||

Atp6ap2
Genomic Map