siRNA Details
| Gene Symbol: Ecd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027475.3 | NP_081751.1 | 2 | 463 | ||
| GenBank mRNA | AK028664.1 | - | 2 | 427 | ||
| AK029169.1 | - | 2 | 395 | |||
| AK043261.1 | - | 2 | 312 | |||
| AK213825.1 | - | 2 | 158 | |||
| BC013470.1 | - | 2 | 412 | 4220504 | ||

Ecd
Genomic Map