siRNA Details
| Gene Symbol: Tmem144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027495.4 | NP_081771.2 | 11, 12 | 1306 | ||
| GenBank mRNA | AK017808.1 | - | 11, 12 | 1325 | ||
| AK079384.1 | - | 11, 12 | 1307 | |||
| BC018493.1 | - | 11, 12 | 1214 | 3593194 | ||

Tmem144
Genomic Map