siRNA Details
| Gene Symbol: Hmha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142701.1 | NP_001136173.1 | 12, 13 | 1661 | ||
| NM_027521.3 | NP_081797.1 | 11, 12 | 1213 | |||
| GenBank mRNA | AK044860.1 | - | 11, 12 | 1175 | ||
| AK134029.1 | - | 5, 6 | 635 | |||
| AK151423.1 | - | 12, 13 | 1605 | |||
| AK159194.1 | - | 12, 13 | 1662 | |||
| AK159474.1 | - | 11, 12 | 1705 | |||
| AK161766.1 | - | 4, 5 | 721 | |||
| AK170262.1 | - | 12, 13 | 1647 | |||
| AK171304.1 | - | 12, 13 | 1647 | |||
| BC053750.1 | - | 11, 12 | 1213 | 6333971 | ||

Hmha1
Genomic Map