siRNA Details
| Gene Symbol: Hmha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142701.1 | NP_001136173.1 | 13 | 1748 | ||
| NM_027521.3 | NP_081797.1 | 12 | 1300 | |||
| GenBank mRNA | AK044860.1 | - | 12 | 1262 | ||
| AK134029.1 | - | 6 | 722 | |||
| AK151423.1 | - | 13 | 1692 | |||
| AK159194.1 | - | 13 | 1749 | |||
| AK159474.1 | - | 12 | 1792 | |||
| AK161766.1 | - | 5 | 808 | |||
| AK170262.1 | - | 13 | 1734 | |||
| AK171304.1 | - | 13 | 1734 | |||
| BC053750.1 | - | 12 | 1300 | 6333971 | ||

Hmha1
Genomic Map