siRNA Details
| Gene Symbol: Hmha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142701.1 | NP_001136173.1 | 13 | 1839 | ||
| NM_027521.3 | NP_081797.1 | 12 | 1391 | |||
| GenBank mRNA | AK044860.1 | - | 12 | 1353 | ||
| AK134029.1 | - | 6 | 813 | |||
| AK151423.1 | - | 13 | 1783 | |||
| AK159194.1 | - | 13 | 1840 | |||
| AK159474.1 | - | 12 | 1883 | |||
| AK161766.1 | - | 5 | 899 | |||
| AK170262.1 | - | 13 | 1825 | |||
| AK171304.1 | - | 13 | 1825 | |||
| BC053750.1 | - | 12 | 1391 | 6333971 | ||

Hmha1
Genomic Map