siRNA Details
| Gene Symbol: Dmrtc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038616.2 | NP_001033705.2 | 5 | 792 | ||
| NM_027591.3 | NP_081867.1 | 7 | 973 | |||
| NM_029378.2 | NP_083654.1 | 5 | 787 | |||
| GenBank mRNA | AK014934.1 | - | 7 | 974 | ||
| DQ316772.1 | - | 7 | 929 | |||
| DQ316773.1 | - | 5 | 787 | |||
| DQ316774.1 | - | 5 | 704 | |||

Dmrtc1a
Genomic Map