siRNA Details
| Gene Symbol: Fam71d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027597.4 | NP_081873.2 | 3, 4 | 471 | ||
| NM_029069.1 | NP_083345.1 | 3, 4 | 476 | |||
| GenBank mRNA | AB473726.1 | - | 1, 2 | 151 | ||
| AK014851.1 | - | 3, 4 | 477 | |||
| AK015802.1 | - | 3, 4 | 324 | |||
| BC051149.1 | - | 3, 4 | 331 | 6707726 | ||

Fam71d
Genomic Map