siRNA Details
| Gene Symbol: 4921504E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027600.4 | NP_081876.2 | 7, 8 | 833 | ||
| GenBank mRNA | AK014808.1 | - | 7, 8 | 834 | ||
| AK014944.1 | - | 7, 8 | 838 | |||
| AK033749.1 | - | 7, 8 | 839 | |||
| BC132142.1 | - | 7, 8 | 823 | 40130419 | ||
| BC137968.1 | - | 7, 8 | 823 | 8860987 | ||

4921504E06Rik
Genomic Map