siRNA Details
| Gene Symbol: Spata1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027617.3 | NP_081893.1 | 3 | 313 | ||
| GenBank mRNA | BC028997.1 | - | 3 | 346 | ||
| BC042647.1 | - | 3 | 346 | |||
| BC055056.1 | - | 3 | 374 | |||
| BC139279.1 | - | 2 | 226 | 8862301 | ||
| BC139280.1 | - | 2 | 226 | 8862302 | ||
| BC171970.1 | - | 2 | 226 | |||

Spata1
Genomic Map