siRNA Details
| Gene Symbol: Spata1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027617.3 | NP_081893.1 | 7 | 793 | ||
| GenBank mRNA | BC139279.1 | - | 6 | 706 | 8862301 | |
| BC139280.1 | - | 6 | 706 | 8862302 | ||
| BC171970.1 | - | 6 | 706 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata1| Gene Symbol: Spata1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027617.3 | NP_081893.1 | 7 | 793 | ||
| GenBank mRNA | BC139279.1 | - | 6 | 706 | 8862301 | |
| BC139280.1 | - | 6 | 706 | 8862302 | ||
| BC171970.1 | - | 6 | 706 | |||