siRNA Details
| Gene Symbol: Sept12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027669.3 | NP_081945.1 | 8 | 905 | ||
| NM_029374.1 | NP_083650.1 | 8 | 905 | |||
| GenBank mRNA | AK006458.1 | - | 8 | 843 | ||
| AK016798.1 | - | 8 | 833 | |||
| AK077091.1 | - | 8 | 906 | |||
| BC115618.1 | - | 8 | 816 | 40102763 | ||
| BC115619.1 | - | 7 | 951 | 40102765 | ||

Sept12
Genomic Map