siRNA Details
| Gene Symbol: Tssk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253888.1 | NP_001240817.1 | 4 | 1043 | ||
| NM_001253889.1 | NP_001240818.1 | 4 | 1190 | |||
| NM_027673.3 | NP_081949.1 | 4 | 1013 | |||
| GenBank mRNA | AK006144.1 | - | 4 | 1165 | ||
| AK016890.1 | - | 4 | 1015 | |||
| AY461664.1 | - | 4 | 905 | |||
| DQ397204.1 | - | 4 | 942 | |||
| DQ397205.1 | - | 4 | 943 | |||
| DQ397206.1 | - | 4 | 1090 | |||
| EF127819.1 | - | 4 | 1015 | |||
| EF127820.1 | - | 4 | 1045 | |||
| EF127821.1 | - | 4 | 1191 | |||
| EF127822.1 | - | 3 | 1458 | |||

Tssk4
Genomic Map