siRNA Details
| Gene Symbol: Daw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027725.3 | NP_082001.2 | 4 | 332 | ||
| GenBank mRNA | AK016201.1 | - | 4 | 306 | ||
| AK016977.1 | - | 4 | 333 | |||
| BC049704.1 | - | 4 | 332 | |||
| BC144994.1 | - | 4 | 309 | |||
| BC145995.1 | - | 4 | 332 | 40131298 | ||
| BU612798.1 | - | 4 | 275 | |||

Daw1
Genomic Map