siRNA Details
| Gene Symbol: Dmrtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027732.2 | NP_082008.1 | 2 | 215 | ||
| GenBank mRNA | AF542046.1 | - | 2 | 212 | ||
| AK017020.1 | - | 2 | 213 | |||
| BC100434.1 | - | 2 | 215 | 30918218 | ||
| BC138203.1 | - | 2 | 146 | 8861222 | ||
| BC145988.1 | - | 2 | 146 | 40131291 | ||

Dmrtc2
Genomic Map