siRNA Details
Gene Symbol: Tchhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027762.3 | NP_082038.2 | 3 | 323 | ||
GenBank mRNA | AA856087.1 | - | 2 | 193 | ||
AK017244.1 | - | 3 | 324 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tchhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027762.3 | NP_082038.2 | 3 | 323 | ||
GenBank mRNA | AA856087.1 | - | 2 | 193 | ||
AK017244.1 | - | 3 | 324 |