siRNA Details
| Gene Symbol: Obox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145708.2 | NP_663754.2 | 4 | 1222 | ||
| GenBank mRNA | BC094912.1 | - | 4 | 1102 | ||
| DV647958.1 | - | 1 | 610 | |||
| Gene Symbol: Obox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027802.2 | NP_082078.1 | 4 | 1130 | ||
| GenBank mRNA | AK018362.1 | - | 4 | 1127 | ||
| AK136061.1 | - | 4 | 1120 | |||

Obox2
Genomic Map