siRNA Details
| Gene Symbol: Lrwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027891.4 | NP_082167.2 | 14 | 1860 | ||
| GenBank mRNA | AK004717.1 | - | 12 | 2822 | ||
| AK084948.1 | - | 14 | 1860 | |||
| AK085327.1 | - | 3 | 1891 | |||
| AK153976.1 | - | 14 | 1831 | |||
| AK154352.1 | - | 14 | 1803 | |||
| AK167020.1 | - | 14 | 1809 | |||
| AK167868.1 | - | 2 | 209 | |||
| AK172708.1 | - | 14 | 1829 | |||
| AK208344.1 | - | 1 | 43 | |||
| BC048397.1 | - | 14 | 1839 | 6396815 | ||

Lrwd1
Genomic Map