siRNA Details
| Gene Symbol: Gtf3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027901.2 | NP_082177.2 | 3 | 730 | ||
| GenBank mRNA | AK004895.1 | - | 3 | 508 | ||
| AK035792.1 | - | 3 | 496 | |||
| AK083585.1 | - | 3 | 731 | |||
| AK083636.1 | - | 2 | 485 | |||
| AK088596.1 | - | 3 | 498 | |||
| AK129033.1 | - | 2 | 480 | |||
| AK138760.1 | - | 499 | ||||
| AK149771.1 | - | 3 | 498 | |||
| AK151300.1 | - | 3 | 495 | |||
| AK152439.1 | - | 3 | 495 | |||
| AK154056.1 | - | 3 | 511 | |||
| AK159938.1 | - | 3 | 497 | |||
| AK181263.1 | - | 1 | 104 | |||
| AK192122.1 | - | 1 | 104 | |||
| AK198852.1 | - | 1 | 104 | |||
| BC034369.1 | - | 3 | 508 | 3994677 | ||
| BC043100.1 | - | 3 | 718 | |||

Gtf3c2
Genomic Map