siRNA Details
| Gene Symbol: Gtf3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027901.2 | NP_082177.2 | 3 | 736 | ||
| GenBank mRNA | AK004895.1 | - | 3 | 514 | ||
| AK035792.1 | - | 3 | 502 | |||
| AK043944.1 | - | 3 | 503 | |||
| AK083585.1 | - | 3 | 737 | |||
| AK083636.1 | - | 2 | 491 | |||
| AK088596.1 | - | 3 | 504 | |||
| AK129033.1 | - | 2 | 486 | |||
| AK138760.1 | - | 505 | ||||
| AK149771.1 | - | 3 | 504 | |||
| AK151300.1 | - | 3 | 501 | |||
| AK152439.1 | - | 3 | 501 | |||
| AK154056.1 | - | 3 | 517 | |||
| AK159938.1 | - | 3 | 503 | |||
| AK181263.1 | - | 1 | 110 | |||
| AK192122.1 | - | 1 | 110 | |||
| AK198852.1 | - | 1 | 110 | |||
| BC034369.1 | - | 3 | 514 | 3994677 | ||
| BC043100.1 | - | 3 | 724 | |||

Gtf3c2
Genomic Map