siRNA Details
| Gene Symbol: Klhdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163729.2 | NP_001157201.2 | 9 | 1262 | ||
| NM_027910.3 | NP_082186.2 | 10 | 1415 | |||
| GenBank mRNA | AB053465.1 | - | 10 | 1286 | ||
| AJ278127.1 | - | 10 | 1282 | |||
| AK004964.1 | - | 10 | 1392 | |||
| AK079082.1 | - | 10 | 1296 | |||
| AK141673.1 | - | 9 | 1247 | |||
| AK170623.1 | - | 10 | 1287 | |||
| BC018154.1 | - | 10 | 1384 | 3983436 | ||

Klhdc3
Genomic Map