siRNA Details
| Gene Symbol: March8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005032.2 | - | 7 | 1160 | ||
| AK152569.1 | - | 1 | 181 | |||
| AK154976.1 | - | 8 | 1352 | |||
| AK165647.1 | - | 7 | 1153 | |||
| BC050908.1 | - | 8 | 1335 | 6416955 | ||
| BC053090.1 | - | 7 | 1420 | 5709246 | ||

March8
Genomic Map