siRNA Details
| Gene Symbol: Osgin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027950.1 | NP_082226.1 | 3, 4 | 382 | ||
| GenBank mRNA | AB221676.1 | - | 2, 3 | 209 | ||
| AK005892.1 | - | 1, 2 | 145 | |||
| BC022135.1 | - | 3, 4 | 382 | 5099665 | ||
| BC110674.1 | - | 3, 4 | 369 | |||
| BC132440.1 | - | 3, 4 | 351 | 40130717 | ||
| BC138192.1 | - | 3, 4 | 351 | 8861211 | ||

Osgin1
Genomic Map