siRNA Details
| Gene Symbol: Cfap52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027963.2 | NP_082239.2 | 8 | 930 | ||
| GenBank mRNA | AK006118.1 | - | 8 | 930 | ||
| AK016839.1 | - | 8 | 941 | |||
| BC118541.1 | - | 8 | 929 | 40058756 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap52| Gene Symbol: Cfap52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027963.2 | NP_082239.2 | 8 | 930 | ||
| GenBank mRNA | AK006118.1 | - | 8 | 930 | ||
| AK016839.1 | - | 8 | 941 | |||
| BC118541.1 | - | 8 | 929 | 40058756 | ||