siRNA Details
| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027992.3 | NP_082268.2 | 9 | 1364 | ||
| GenBank mRNA | AK009646.1 | - | 9 | 1265 | ||
| AK148477.1 | - | 9 | 1288 | |||
| BC022695.1 | - | 4 | 551 | |||
| BC050246.1 | - | 9 | 1281 | 6313498 | ||

Tmem106b
Genomic Map