siRNA Details
| Gene Symbol: Dus4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028002.2 | NP_082278.1 | 3, 4 | 414 | ||
| GenBank mRNA | AK010138.1 | - | 3, 4 | 415 | ||
| BC109351.2 | - | 3, 4 | 349 | 40056894 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dus4l| Gene Symbol: Dus4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028002.2 | NP_082278.1 | 3, 4 | 414 | ||
| GenBank mRNA | AK010138.1 | - | 3, 4 | 415 | ||
| BC109351.2 | - | 3, 4 | 349 | 40056894 | ||