siRNA Details
| Gene Symbol: Rpap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028003.2 | NP_082279.1 | 10 | 1181 | ||
| GenBank mRNA | AK009765.1 | - | 10 | 1182 | ||
| AK169097.1 | - | 10 | 1134 | |||
| BC004046.1 | - | 10 | 1184 | 3496407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpap3| Gene Symbol: Rpap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028003.2 | NP_082279.1 | 10 | 1181 | ||
| GenBank mRNA | AK009765.1 | - | 10 | 1182 | ||
| AK169097.1 | - | 10 | 1134 | |||
| BC004046.1 | - | 10 | 1184 | 3496407 | ||