siRNA Details
| Gene Symbol: NEDD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142393.1 | NP_001135865.1 | 3 | 760 | ||
| NM_006403.3 | NP_006394.1 | 2 | 565 | |||
| NM_182966.3 | NP_892011.2 | 2 | 565 | |||
| NR_073131.1 | 4 | 854 | ||||
| GenBank mRNA | AK292682.1 | - | 3 | 760 | ||
| BC020686.1 | - | 2 | 536 | |||
| BC040207.1 | - | 2 | 551 | 4820745 | ||
| BX648041.1 | - | 4 | 822 | |||
| DA660812.1 | - | 2 | 567 | |||
| L43821.1 | - | 2 | 561 | |||
| U64317.1 | - | 2 | 536 | |||

NEDD9
Genomic Map