siRNA Details
| Gene Symbol: 2310067B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028014.3 | NP_082290.2 | 26, 27 | 3654 | ||
| GenBank mRNA | AK047622.1 | - | 12, 13 | 1788 | ||
| BC007157.1 | - | 13, 14 | 1900 | |||
| BC030865.1 | - | 11, 12 | 1421 | |||
| BC053088.1 | - | 26, 27 | 3569 | 5709104 | ||

2310067B10Rik
Genomic Map