siRNA Details
| Gene Symbol: Cers5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028015.2 | NP_082291.1 | 1, 2 | 339 | ||
| GenBank mRNA | AK010241.1 | - | 1, 2 | 267 | ||
| AK075694.1 | - | 1, 2 | 259 | |||
| AK145589.1 | - | 1, 2 | 247 | |||
| AK145762.1 | - | 1, 2 | 261 | |||
| AK146220.1 | - | 1, 2 | 261 | |||
| AK146417.1 | - | 1, 2 | 230 | |||
| AK219835.1 | - | 1, 2 | 49 | |||
| AY029533.1 | - | 1, 2 | 259 | |||
| BC043059.1 | - | 286 | 6419704 | |||
| BC046797.1 | - | 1, 2 | 339 | 6403129 | ||

Cers5
Genomic Map